R&D Systems
Manufacturer Code:BAF977
Catalog # BAF977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant rat E-Selectin/CD62E Trp22-Pro494 Accession # P98105 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD62 antigen-like family member E; CD62 antigen-like family member E CD62E CD62E antigen ELAM ELAM1 ELAM-1 ELAM1E-selectin endothelial adhesion molecule 1 Endothelial leukocyte adhesion molecule 1 ESEL LECAM2 leukocyte endothelial cell adhesion molecule 2 Leukocyte-endothelial cell adhesion molecule 2 SELE selectin E; CD62E; E-selectin; ELAM-1; endothelial adhesion molecule 1; Endothelial leukocyte adhesion molecule 1; LECAM2; leukocyte endothelial cell adhesion molecule 2; Leukocyte-endothelial cell adhesion molecule 2
Gene Aliases: CD62E; ELAM; ELAM1; ESEL; LECAM2; SELE
UniProt ID: (Human) P16581
Entrez Gene ID: (Human) 6401
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.