Proteintech
Manufacturer Code:127151AP150UL
Catalog # 50-172-8198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Dystrophin Fusion Protein Ag3408 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apodystrophin-3; apodystrophin-I; BMD CMD3B DMD DXS142 DXS164 DXS206 DXS230 DXS239 DXS268 DXS269 DXS270 DXS272 dystrophin; Dystrophin; dystrophin Dp40; dystrophin Dp71a; dystrophin Dp71ab; dystrophin Dp71b; dystrophin, muscular dystrophy; X-linked muscular dystrophy
Gene Aliases: BMD; CMD3B; DMD; DNADMD1; Dp427; Dp71; DXS142; DXS164; DXS206; DXS230; DXS239; DXS268; DXS269; DXS270; DXS272; DXSmh7; DXSmh9; dys; mdx; MRX85; pke
UniProt ID: (Human) P11532, (Mouse) P11531, (Rat) P11530
Entrez Gene ID: (Human) 1756, (Mouse) 13405, (Rat) 24907
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.