Proteintech
Manufacturer Code:667351IG150UL
Catalog # 50-173-7147
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B1G12 |
Immunogen | Dystroglycan Fusion Protein Ag27222 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 156DAG A3a AGRNR Alpha DG Beta dystroglycan Beta DG DAG DAG1 Dystroglycan; Alpha-DG; Alpha-dystroglycan; Beta-DG; Beta-dystroglycan; Dystroglycan; dystroglycan 1 (dystrophin-associated glycoprotein 1); dystrophin associated glycoprotein 1; Dystrophin-associated glycoprotein 1
Gene Aliases: 156DAG; A3a; AGRNR; D9Wsu13e; DAG; Dag-1; DAG1; DG; Dp427; Dp71; MDDGA9; MDDGC7; MDDGC9
UniProt ID: (Human) Q14118, (Mouse) Q62165
Entrez Gene ID: (Human) 1605, (Pig) 100620392, (Mouse) 13138, (Rat) 114489
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.