R&D Systems
Manufacturer Code:AF6868SP
Catalog # AF6868SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Dystroglycan Gln28-Val749 Accession # Q14118 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: A3a AGRNR Dag-1 DAG1 DAG156DAG dystroglycan dystroglycan 1 (dystrophin-associated glycoprotein 1) Dystrophin-associated glycoprotein 1; Alpha-DG; Alpha-dystroglycan; Beta-DG; Beta-dystroglycan; Dystroglycan; dystroglycan 1 (dystrophin-associated glycoprotein 1); Dystrophin-associated glycoprotein 1
Gene Aliases: 156DAG; A3a; AGRNR; DAG; DAG1; MDDGA9; MDDGC7; MDDGC9
UniProt ID: (Human) Q14118
Entrez Gene ID: (Human) 1605
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.