Novus Biologicals
Manufacturer Code:NBP266814
Catalog # NBP266814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JF05-54 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Dysferlin; dysferlin dysferlin limb girdle muscular dystrophy 2B (autosomal recessive) dystrophy-associated fer-1-like 1 FER1L1Dystrophy-associated fer-1-like protein Fer-1-like protein 1 FLJ00175 FLJ90168 LGMD2B; dystrophy-associated fer-1-like 1; Dystrophy-associated fer-1-like protein; fer-1-like family member 1; Fer-1-like protein 1; limb girdle muscular dystrophy 2B (autosomal recessive)
Gene Aliases: DYSF; FER1L1; LGMD2B; MMD1
UniProt ID: (Human) O75923
Entrez Gene ID: (Human) 8291
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.