Invitrogen
Manufacturer Code:PA553546
Catalog # PIPA553546
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Dysferlin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dysferlin; dystrophy-associated fer-1-like 1; Dystrophy-associated fer-1-like protein; fer-1-like family member 1; Fer-1-like protein 1; FER1L1 LGMD2B MMD1 dysferlin dystrophy-associated fer-1-like 1 dystrophy-associated fer-1-like protein fer-1-like protein 1; limb girdle muscular dystrophy 2B (autosomal recessive)
Gene Aliases: DYSF; FER1L1; LGMD2B; MMD1
UniProt ID: (Human) O75923
Entrez Gene ID: (Human) 8291
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.