Proteintech
Manufacturer Code:111321AP150UL
Catalog # 50-172-6904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Dysbindin Fusion Protein Ag1613 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 8; Biogenesis of lysosome-related organelles complex 1 subunit 8; BLOC-1 subunit 8; DBND DTNBP1 DTNBP1a DTNBP1b Dysbindin Dysbindin 1 dystrobrevin binding protein 1 HPS7 HPS7 protein My031 SDY; distrobrevin binding protein 1; Dysbindin; Dysbindin-1; Dystrobrevin-binding protein 1; Hermansky-Pudlak syndrome 7 protein; Hermansky-Pudlak syndrome 7 protein homolog; HPS7 protein; HPS7 protein homolog; sandy
Gene Aliases: 5430437B18Rik; AW048963; BLOC1S8; DBND; DTNBP1; dysbindin; HPS7; My031; SDY
UniProt ID: (Human) Q96EV8, (Mouse) Q91WZ8, (Rat) Q5M834
Entrez Gene ID: (Human) 84062, (Mouse) 94245, (Rat) 641528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.