Novus Biologicals
Manufacturer Code:NBP292214100UL
Catalog # NB026319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-63 of human DYNLRB1 (NP_001268656.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BITH Bithoraxoid-like protein DNCL2Acytoplasmic dynein light chain 2A DNLC2ABLP Dynein light chain 2A cytoplasmic dynein light chain roadblock-type 1 dynein cytoplasmic light polypeptide 2A dynein light chain roadblock-type 1 dynein-associated protein HKM23 Dynein-associated protein Km23 roadblock domain containing 1 Roadblock domain-containing protein 1 ROBL/LC7-like 1 ROBLD1Roadblock-1; Bithoraxoid-like protein; BLP; Dynein light chain 2A, cytoplasmic; Dynein light chain roadblock-type 1; dynein, cytoplasmic, light polypeptide 2A; dynein, light chain, roadblock-type 1; Dynein-associated protein Km23; Roadblock domain-containing protein 1; ROBL/LC7-like 1
Gene Aliases: BITH; BLP; DNCL2A; DNLC2A; DYNLRB1; HSPC162; ROBLD1
UniProt ID: (Human) Q9NP97
Entrez Gene ID: (Human) 83658
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.