Novus Biologicals
Manufacturer Code:NBP29213720UL
Catalog # NB026167
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-89 of human DYNLL2 (NP_542408.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 8 kDa dynein light chain b; DLC2 DLC8b DNCL1B dynein light chain 2 cytoplasmic Dynein light chain LC8-type 28 kDa dynein light chain b dynein light chain LC8-type 2 MGC17810 radial spoke 22 homolog RSPH22; DLC8b; Dynein light chain 2, cytoplasmic; Dynein light chain LC8-type 2; dynein, light chain, LC8-type 2; radial spoke 22 homolog
Gene Aliases: DLC2; DNCL1B; DYNLL2; RSPH22
UniProt ID: (Human) Q96FJ2
Entrez Gene ID: (Human) 140735
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.