Proteintech
Manufacturer Code:113611AP150UL
Catalog # 50-172-7092
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Dynamitin (1-401 AA) Fusion Protein Ag1902 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 50 kDa dynein-associated polypeptide; DCTN 50 DCTN2 DCTN50 dynactin 2 dynactin 2 (p50) Dynactin subunit 2 DYNAMITIN p50 dynamitin RBP50; DCTN-50; dynactin 2 (p50); Dynactin complex 50 kDa subunit; Dynactin subunit 2; epididymis secretory protein Li 77; GMP23-48K; Growth cone membrane protein 23-48K; p50 dynamitin
Gene Aliases: 2310042E05Rik; C130077D06Rik; C87049; DCTN-50; DCTN2; DCTN50; DYNAMITIN; GMP23-48K; HEL-S-77; p50; RBP50
UniProt ID: (Human) Q13561, (Rat) Q6AYH5, (Mouse) Q99KJ8
Entrez Gene ID: (Human) 10540, (Rat) 299850, (Mouse) 69654
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.