Invitrogen
Manufacturer Code:MA537890
Catalog # PIMA537890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2428 |
Immunogen | Recombinant protein of human Dynamin 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 9630020E24Rik AW061159 B230343F03Rik C530045C17 DNM3 Dyn3 Dyna III dynamin 3 dynamin family member Dynamin III dynamin testicular dynamin-2 Dynamin-3 Kiaa0820 mKIAA0820 T-dynamin testicular dynamin; Dyna III; dynamin family member; Dynamin III; Dynamin, testicular; Dynamin-3; T-dynamin; testicular dynamin
Gene Aliases: 9630020E24Rik; AW061159; B230343F03Rik; C530045C17; DNM3; Dyna III; KIAA0820; mKIAA0820
UniProt ID: (Human) Q9UQ16, (Mouse) Q8BZ98
Entrez Gene ID: (Human) 26052, (Mouse) 103967
Molecular Function:
G-protein
cytoskeletal protein
enzyme modulator
hydrolase
microtubule family cytoskeletal protein
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.