Invitrogen
Manufacturer Code:PA590253
Catalog # PIPA590253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 410-555 of human DNM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dyna III; Dynamin III; Dynamin, testicular; Dynamin-3; T-dynamin; testicular dynamin
Gene Aliases: 9630020E24Rik; AW061159; B230343F03Rik; C530045C17; Dnm3; Dyn3; dynamin-2; Kiaa0820; mKIAA0820
UniProt ID: (Mouse) Q8BZ98, (Rat) Q08877
Entrez Gene ID: (Mouse) 103967, (Rat) 171574
Molecular Function:
G-protein
cytoskeletal protein
enzyme modulator
hydrolase
microtubule family cytoskeletal protein
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.