Invitrogen
Manufacturer Code:PA5116413
Catalog # PIPA5116413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 557-584 amino acids from the Central region of human DNM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9630020E24Rik AW061159 B230343F03Rik C530045C17 DNM3 Dyn3 Dyna III dynamin 3 dynamin family member Dynamin III dynamin testicular dynamin-2 Dynamin-3 Kiaa0820 mKIAA0820 T-dynamin testicular dynamin; dynamin family member; Dynamin, testicular; Dynamin-3; T-dynamin
Gene Aliases: DNM3; Dyna III; KIAA0820
UniProt ID: (Human) Q9UQ16
Entrez Gene ID: (Human) 26052
Molecular Function:
G-protein
cytoskeletal protein
enzyme modulator
hydrolase
microtubule family cytoskeletal protein
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.