Novus Biologicals
Manufacturer Code:NBP29262720UL
Catalog # NB027125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 410-555 of human DNM3 (NP_001265181.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dyna III dynamin 3 dynamin family member dynamin-3 EC 3.6.5.5 MGC70433 T-dynamin testicular; dynamin family member; Dynamin, testicular; Dynamin-3; T-dynamin
Gene Aliases: DNM3; Dyna III; KIAA0820
UniProt ID: (Human) Q9UQ16
Entrez Gene ID: (Human) 26052
Molecular Function:
G-protein
cytoskeletal protein
enzyme modulator
hydrolase
microtubule family cytoskeletal protein
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.