Novus Biologicals
Manufacturer Code:NBP261757
Catalog # NBP2-61757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F5F3 |
Immunogen | Purified recombinant fragment of human Dynamin-2 (AA: 520-744) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: CMTDI1CMT2M CMTDIBdynamin II DI-CMTB DYN2EC 3.6.5.5 dynamin 2 dynamin-2 DYNII; dynamin II; Dynamin-2
Gene Aliases: CMT2M; CMTDI1; CMTDIB; DI-CMTB; DNM2; DYN2; DYNII; LCCS5
UniProt ID: (Human) P50570
Entrez Gene ID: (Human) 1785
Molecular Function: G-protein cytoskeletal protein enzyme modulator hydrolase microtubule family cytoskeletal protein small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.