Invitrogen
Manufacturer Code:MA534957
Catalog # PIMA534957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | A1A12 |
Immunogen | Recombinant protein within human Dynamin 1 aa 500-800. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AI838169 B-dynamin BOS_12077 D100 DNM DNM1 Dyn1 Dynamin BREDNM19 Dynamin brain Dynamin1 Dynamin-1 EC 3.6.5.5 EIEE31 Ftfl Kiaa4093 mKIAA4093 Neuronal dynamin OTTHUMP00000215500; Dynamin-1
Gene Aliases: DNM; DNM1; EIEE31
UniProt ID: (Human) Q05193
Entrez Gene ID: (Human) 1759
Molecular Function: G-protein cytoskeletal protein enzyme modulator hydrolase microtubule family cytoskeletal protein small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.