Invitrogen
Manufacturer Code:PA544893
Catalog # PIPA544893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human DCTN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 50 kDa dynein-associated polypeptide; DCTN-50; dynactin 2 (p50); Dynactin 2 (p50) DCTN50 DYNAMITIN RBP50; Dynactin complex 50 kDa subunit; Dynactin subunit 2; epididymis secretory protein Li 77; GMP23-48K; Growth cone membrane protein 23-48K; p50 dynamitin; p50/dynamitin
Gene Aliases: 2310042E05Rik; C130077D06Rik; C87049; DCTN-50; DCTN2; DCTN50; DYNAMITIN; GMP23-48K; HEL-S-77; p50; RBP50; zgc:63867
UniProt ID: (Bovine) Q3ZCF0, (Human) Q13561, (Rat) Q6AYH5, (Mouse) Q99KJ8, (Zebrafish) Q7T3H1
Entrez Gene ID: (Bovine) 527201, (Dog) 474415, (Rabbit) 100339187, (Human) 10540, (Horse) 100050464, (Rat) 299850, (Mouse) 69654, (Guinea pig) 100721002, (Zebrafish) 394141
Molecular Function:
cytoskeletal protein
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.