Abnova
Manufacturer Code:PAB27670
Catalog # 89-129-184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of mouse Dvl1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Dvl1.
For Research Use Only
Protein Aliases: Dishevelled-1; DSH homolog 1; Dvl; KIAA4029; mKIAA4029; Segment polarity protein dishevelled homolog DVL-1
Gene Aliases: Dvl; Dvl1; mKIAA4029
UniProt ID: (Mouse) P51141
Entrez Gene ID: (Mouse) 13542
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.