Invitrogen
Manufacturer Code:PA512076
Catalog # PIPA512076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 20-50 amino acids from the N-terminal region of human Dsk2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chap1; DSK2 homolog; Nedd4 binding protein 4; PLIC-2; Protein linking IAP with cytoskeleton 2; Ubiquilin 2 Protein linking IAP with cytoskeleton-2 PLIC-2 hPLIC-2 Ubiquitin-like product Chap1/Dsk2 DSK2 homolog Chap1 HRIHFB2157; Ubiquilin-2; Ubiquitin-like product Chap1/Dsk2
Gene Aliases: ALS15; CHAP1; DSK2; HRIHFB2157; N4BP4; PLIC2; UBQLN2
UniProt ID: (Human) Q9UHD9
Entrez Gene ID: (Human) 29978
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.