Invitrogen
Manufacturer Code:MA535315
Catalog # PIMA535315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0077 |
Immunogen | A synthesized peptide derived from human Drosha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 1110013A17Rik AI874853 DROSHA Etohi2 HSA242976 MGC115770 OTTHUMP00000220910 OTTHUMP00000220911 OTTHUMP00000220912 OTTHUMP00000220913 p241 Protein Drosha RANSE3L RGD1307626 Ribonuclease 3 Ribonuclease III Rn3 RNase III RNASE3L RNASEN; drosha, double-stranded RNA-specific endoribonuclease; drosha, ribonuclease type III; nuclear RNase III Drosha; p241; Protein Drosha; putative protein p241 which interacts with transcription factor Sp1; putative ribonuclease III; Ribonuclease 3; Ribonuclease III; ribonuclease III, nuclear; ribonuclease type III, nuclear; RNase III
Gene Aliases: DROSHA; ETOHI2; HSA242976; RANSE3L; RN3; RNASE3L; RNASEN
UniProt ID: (Human) Q9NRR4
Entrez Gene ID: (Human) 29102
Molecular Function: RNA binding protein endoribonuclease nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.