Invitrogen
Manufacturer Code:PA5103162
Catalog # PIPA5103162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RNase III Drosha corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110013A17Rik AI874853 Drosha drosha ribonuclease III drosha double-stranded RNA-specific endoribonuclease drosha ribonuclease type III ethanol induced 2 Etohi2 HSA242976 MGC115770 nuclear RNase III Drosha OTTHUMP00000220910 OTTHUMP00000220911 OTTHUMP00000220912 OTTHUMP00000220913 p241 Protein Drosha putative protein p241 which interacts with transcription factor Sp1 putative ribonuclease III RANSE3L RGD1307626 Ribonuclease 3 Ribonuclease III ribonuclease III nuclear ribonuclease type III nuclear RN3 RNase III RNASE3L Rnasen; drosha, double-stranded RNA-specific endoribonuclease; drosha, ribonuclease type III; ethanol induced 2; nuclear RNase III Drosha; p241; Protein Drosha; putative protein p241 which interacts with transcription factor Sp1; putative ribonuclease III; Ribonuclease 3; Ribonuclease III; ribonuclease III, nuclear; ribonuclease type III, nuclear; RNase III
Gene Aliases: 1110013A17Rik; AI874853; DROSHA; ETOHI2; HSA242976; RANSE3L; RN3; RNASE3L; RNASEN
UniProt ID: (Human) Q9NRR4, (Mouse) Q5HZJ0
Entrez Gene ID: (Human) 29102, (Mouse) 14000
Molecular Function:
RNA binding protein
endoribonuclease
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.