Novus Biologicals
Manufacturer Code:NBP15713920UL
Catalog # NBP15713920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RNASEN(ribonuclease type III nuclear) The peptide sequence was selected from the middle region of RNASEN. Peptide sequence AAMDALEKYNFPQMAHQKRFIERKYRQELKEMRWEREHQEREPDETEDIK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: drosha double-stranded RNA-specific endoribonuclease drosha ribonuclease type III EC 3.1.26.3 Etohi2 nuclear RNase III Drosha p241 Protein Drosha putative protein p241 which interacts with transcription factor Sp1 putative ribonuclease III RANSE3L ribonuclease 3 Ribonuclease III ribonuclease III nuclear RN3nuclear RNase III; drosha, double-stranded RNA-specific endoribonuclease; drosha, ribonuclease type III; nuclear RNase III Drosha; p241; Protein Drosha; putative protein p241 which interacts with transcription factor Sp1; putative ribonuclease III; Ribonuclease 3; Ribonuclease III; ribonuclease III, nuclear; ribonuclease type III, nuclear; RNase III
Gene Aliases: DROSHA; ETOHI2; HSA242976; RANSE3L; RN3; RNASE3L; RNASEN
UniProt ID: (Human) Q9NRR4
Entrez Gene ID: (Human) 29102
Molecular Function: RNA binding protein endoribonuclease nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.