Proteintech
Manufacturer Code:257701AP150UL
Catalog # 50-173-5067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Drebrin Fusion Protein Ag22634 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D0S117E DBN1 Drebrin drebrin 1; Developmentally-regulated brain protein; Drebrin; drebrin A; drebrin E; drebrin E2; drebrin-1
Gene Aliases: D0S117E; DBN1; Drba
UniProt ID: (Human) Q16643, (Mouse) Q9QXS6
Entrez Gene ID: (Human) 1627, (Mouse) 56320
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.