Novus Biologicals
Manufacturer Code:NBP297115AF594
Catalog # NB042026
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Draxin/C1orf187 (NP_940947.3 Met1-Val349). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGPA3119 C1orf187 chromosome 1 open reading frame 187 dorsal inhibitory axon guidance protein Dorsal repulsive axon guidance protein draxin FLJ34999 MGC117222 neucrin UNQ3119; Dorsal inhibitory axon guidance protein; Dorsal repulsive axon guidance protein; Draxin; Neucrin; neural tissue-specific cysteine-rich protein
Gene Aliases: AGPA3119; C1orf187; DRAXIN; neucrin; PSEC0258; UNQ3119; UNQ3119/PRO10268
UniProt ID: (Human) Q8NBI3
Entrez Gene ID: (Human) 374946
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.