Invitrogen
Manufacturer Code:PA572708
Catalog # PIPA572708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Neucrin antibody for a 17 amino acid synthetic peptide near the carboxy terminus of human Neucrin. The immunogen is within the last 50 amino acids of NEUCRIN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf187 Neucrin AGPA3119 C1orf187 chromosome 1 open reading frame 187 dorsal inhibitory axon guidance protein Dorsal repulsive axon guidance protein DRAXIN draxin FLJ34999 MGC117222 neucrin Neucrin UNQ3119; Dorsal inhibitory axon guidance protein; Dorsal repulsive axon guidance protein; Draxin; Neucrin; neural tissue-specific cysteine-rich protein
Gene Aliases: 2610109H07Rik; AGPA3119; C1orf187; DRAXIN; neucrin; PSEC0258; UNQ3119; UNQ3119/PRO10268
UniProt ID: (Human) Q8NBI3, (Mouse) Q6PAL1
Entrez Gene ID: (Human) 374946, (Rat) 691317, (Mouse) 70433
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.