Invitrogen
Manufacturer Code:PA519028
Catalog # PIPA519028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (PQGRSSRQDKYS) corresponding to the N-terminus amino acids of Dpm1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: dolichol-phosphate (beta-D) mannosyltransferase 1; dolichol-phosphate mannosyltransferase subunit 1; dolichyl-phosphate mannosyltransferase 1; DPM1 dolichyl-phosphate mannosyltransferase polypeptide 1 catalytic subunit RP23-391M18.1 AI118379 AI194292 OTTMUSP00000017164 dolichol monophosphate mannose synthase dolichyl-phosphate mannosyltransferase polypeptide 1
Entrez Gene ID: (Rat) 296394
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.