Novus Biologicals
Manufacturer Code:NBP192684SS
Catalog # NBP192684SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 30 |
Immunogen | Recombinant full length human Lis-A isoform of Doublecortin purified from E. coli. [UniProt# O43602] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: doublecortex; doublecortin Doublin FLJ51296 Lissencephalin-X Lis-X X-linked (doublecortin); Doublin; Lis-X; Lissencephalin-X; Neuronal migration protein doublecortin
Gene Aliases: DBCN; DC; DCX; LISX; SCLH; XLIS
UniProt ID: (Human) O43602
Entrez Gene ID: (Human) 1641
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.