Invitrogen
Manufacturer Code:PA517428
Catalog # PIPA517428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human doublecortin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: doublecortex; Doublin; Lis-X; Lissencephalin-X; lissencephaly, X-linked (doublecortin); Neuronal migration protein doublecortin; OTTHUMP00000023860 OTTHUMP00000216316 doublecortex doublin lis-X lissencephalin-X neuronal migration protein doublecortin
Gene Aliases: DBCN; Dbct; DC; Dcn; DCX; LISX; SCLH; XLIS
UniProt ID: (Human) O43602, (Mouse) O88809
Entrez Gene ID: (Human) 1641, (Rat) 84394, (Mouse) 13193
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.