Novus Biologicals
Manufacturer Code:NBP257836
Catalog # NBP257836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LINRFNNEDVCTCPQASVSQQFTSVPWNSFNRDVLKALYSFAPISMHCNKSSAVRFQGEWNLQPLPKVISTLEEPTPQCPTSQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DBM dopamine beta-hydroxylase dopamine beta-hydroxylase (dopamine beta-monooxygenase) Dopamine beta-monooxygenase EC 1.14.17.1; Dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); Dopamine beta-monooxygenase; Soluble dopamine beta-hydroxylase
Gene Aliases: DBH; DBM
UniProt ID: (Human) P09172
Entrez Gene ID: (Human) 1621
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.