R&D Systems
Manufacturer Code:AF7376SP
Catalog # AF7376SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Dopamine beta-Hydroxylase Leu37-Gly617 Accession # P09172 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: DBH DBM dopamine beta-hydroxylase Dopamine betaHydroxylase dopamine beta-hydroxylase (dopamine beta-monooxygenase) Dopamine beta-monooxygenase DOPBHY EC 1.14.17.1; Dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); Dopamine beta-monooxygenase; Soluble dopamine beta-hydroxylase
Gene Aliases: DBH; DBM
UniProt ID: (Human) P09172
Entrez Gene ID: (Human) 1621
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.