Invitrogen
Manufacturer Code:PA118314
Catalog # 01-670-567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Inhibition Assays (IA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Radioimmune Assays (RIA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native enzyme prepared from bovine adrenal glands. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dopamine beta hydroxylase (dopamine beta-monooxygenase); Dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase precursor (EC 1.14.17.1); Dopamine beta-monooxygenase; dopamine beta-monooxygenase precursor (EC 1.14.17.1); mixed function oxidase; Soluble dopamine beta-hydroxylase
Gene Aliases: DBH; DBM; DOPBHY
UniProt ID: (Human) P09172, (Bovine) P15101, (Rat) Q05754
Entrez Gene ID: (Human) 1621, (Bovine) 280758, (Rat) 25699, (Guinea pig) 100727873
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.