Invitrogen
Manufacturer Code:MA535010
Catalog # PIMA535010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL3123 |
Immunogen | Recombinant Protein corresonding to Human Dopamine Transporter |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DA transporter; DA transporter DAT Dat1 dopamine transporter dopamine transporter 1 PKDYS SLC6A3 sodium-dependent dopamine transporter solute carrier family 6 (neurotransmitter transporter) member 3 solute carrier family 6 (neurotransmitter transporter dopamine) member 3 solute carrier family 6 member 3 solute carrier family 6 member 3; DAT; dopamine transporter 1; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; Solute carrier family 6 member 3; solute carrier family 6, member 3
Gene Aliases: DAT; DAT1; PKDYS; SLC6A3
UniProt ID: (Human) Q01959, (Rat) P23977, (Mouse) Q61327
Entrez Gene ID: (Human) 6531, (Rat) 24898, (Mouse) 13162
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.