Invitrogen
Manufacturer Code:MA518079
Catalog # PIMA518079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6V-23-23 |
Immunogen | Synthetic amino acid N-terminal peptide of rat dopamine transporter |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DA transporter; DAT; dopamine transporter 1; Sodium-dependent dopamine transporter; Sodium-dependent dopamine transporter Solute carrier family 6 member 3 DA transporter DAT DAT1; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; Solute carrier family 6 member 3; solute carrier family 6, member 3
Gene Aliases: DAT; Dat1; Slc6a3
UniProt ID: (Rat) P23977, (Mouse) Q61327
Entrez Gene ID: (Rat) 24898, (Mouse) 13162
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.