Novus Biologicals
Manufacturer Code:NBP213939
Catalog # NBP213939
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: PRSNDTAYPGQLALYQQLAQGNAVG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: D(1B) dopamine receptor; D(1B) dopamine receptor D(5) dopamine receptor D1beta dopamine receptor Dopamine D5 receptor dopamine receptor D1B dopamine receptor D5 DRD1BDBDR DRD1L2MGC10601; D(5) dopamine receptor; D1beta dopamine receptor; Dopamine D5 receptor; dopamine receptor D1B
Gene Aliases: DBDR; DRD1B; DRD1L2; DRD5
UniProt ID: (Human) P21918
Entrez Gene ID: (Human) 1816
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.