Novus Biologicals
Manufacturer Code:NBP29217820UL
Catalog # NB026248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DRD3 (NP_000787.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(3) dopamine receptor; D(3) dopamine receptor D3DR Dopamine D3 receptor dopamine receptor D3 essential tremor 1 ETM1 FET1 MGC149204 MGC149205; Dopamine D3 receptor; essential tremor 1
Gene Aliases: D3DR; DRD3; ETM1; FET1
UniProt ID: (Human) P35462
Entrez Gene ID: (Human) 1814
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.