Novus Biologicals
Manufacturer Code:NBP303065
Catalog # NB094679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human Dopamine D2R/DRD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D(2) dopamine receptor; D(2) dopamine receptor D2DR D2R Dopamine D2 receptor dopamine receptor D2 dopamine receptor D2 isoform seven transmembrane helix receptor; Dopamine D2 receptor; dopamine receptor D2 isoform; seven transmembrane helix receptor
Gene Aliases: D2DR; D2R; DRD2
UniProt ID: (Human) P14416
Entrez Gene ID: (Human) 1813
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.