Novus Biologicals
Manufacturer Code:NB11060017AF647
Catalog # NB11060017Y
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry - Free Floating (IHC (Free)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SG2-D1a |
Immunogen | Recombinant rat Dopamine Receptor D1 protein near the C-terminus. [Swiss-Prot# P18901] |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: D(1A) dopamine receptor; D(1A) dopamine receptor D1DR DADR Dopamine D1 receptor dopamine receptor D1 DRD1A; Dopamine D1 receptor
Gene Aliases: DADR; DRD1; DRD1A
UniProt ID: (Human) P21728
Entrez Gene ID: (Human) 1812
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.