Novus Biologicals
Manufacturer Code:NBP289567DL594
Catalog # NB067330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 40 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Dopa Decarboxylase/DDC (NP_000781.1 Met 1-Glu 480). |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AADC; AADCaromatic-L-amino-acid decarboxylase DOPA decarboxylase dopa decarboxylase (aromatic L-amino acid decarboxylase) EC 4.1.1 EC 4.1.1.28; aromatic L-amino acid decarboxylase; Aromatic-L-amino-acid decarboxylase; DDC; DOPA decarboxylase; dopa decarboxylase (aromatic L-amino acid decarboxylase)
Gene Aliases: AADC; DDC
UniProt ID: (Human) P20711
Entrez Gene ID: (Human) 1644
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.