Abnova
Manufacturer Code:PAB15726
Catalog # 89-117-791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 254 mouse Dock4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Dock4.
For Research Use Only
Protein Aliases: 5330406C03; 6330411N01Rik; AF263288; C030023J22; Dedicator of cytokinesis protein 4; mKIAA0716
Gene Aliases: 5330406C03; 6330411N01Rik; AF263288; C030023J22; Dock4; Kiaa0716; mKIAA0716
UniProt ID: (Mouse) P59764
Entrez Gene ID: (Mouse) 238130
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.