Novus Biologicals
Manufacturer Code:NBP292029100UL
Catalog # NB025951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 192-391 of human TRDMT1 (NP_004403.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DMNT2 DNA (cytosine-5-)-methyltransferase 2 DNA (cytosine-5)-methyltransferase-like protein 2 DNA methyltransferase homolog HsaIIP DNA methyltransferase-2 DNA MTase homolog HsaIIP Dnmt2 EC 2.1.1 EC 2.1.1.29 M.HsaIIP MHSAIIP PuMet RNMT1 tRNA (cytosine-5-)-methyltransferase tRNA aspartic acid methyltransferase 1; DNA (cytosine-5)-methyltransferase-like protein 2; DNA cytosine-5 methyltransferase 2; DNA methyltransferase homolog HsaIIP; DNA methyltransferase-2; DNA MTase homolog HsaIIP; Dnmt2; PuMet; tRNA (cytosine(38)-C(5))-methyltransferase; tRNA (cytosine-5-)-methyltransferase
Gene Aliases: DMNT2; DNMT2; MHSAIIP; PUMET; RNMT1; TRDMT1
UniProt ID: (Human) O14717
Entrez Gene ID: (Human) 1787
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.