Abnova
Manufacturer Code:MAB16522
Catalog # 89-999-404
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | G28 |
Immunogen | Recombinant protein corresponding to amino acids 1-104 of rat Dlg1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Disks large homolog 1; Dlgh1; Drosophila discs-large tumor suppressor homologue (synapse associated protein); SAP-97; SAP97; Synapse-associated protein 97
Gene Aliases: Dlg1; Dlgh1; SAP97
UniProt ID: (Rat) Q62696
Entrez Gene ID: (Rat) 25252
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.