Novus Biologicals
Manufacturer Code:NBP290496FR
Catalog # NB074274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 19 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse Dkk-3 (NP_056629.1 Met 1-Ile 349). |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: dickkopf (Xenopus laevis) homolog 3 dickkopf 3 dickkopf homolog 3 (Xenopus laevis) Dickkopf-3 dickkopf-related protein 3 Dkk-3 hDkk-3 regulated in glioma REICdkk-3 RIG RIG-like 5-6 RIG-like 7-1; dickkopf 3 homolog; dickkopf homolog 3; Dickkopf-3; Dickkopf-related protein 3; dkk-3; hDkk-3; regulated in glioma; RIG-like 5-6; RIG-like 7-1
Gene Aliases: DKK3; REIC; RIG; UNQ258/PRO295
UniProt ID: (Human) Q9UBP4
Entrez Gene ID: (Human) 27122
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.