Novus Biologicals
Manufacturer Code:NB1001450
Catalog # NB1001450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence CDHHQASNSSRLHT corresponding to C-Terminus according to NP_036374.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: dickkopf (Xenopus laevis) homolog 1 dickkopf homolog 1 (Xenopus laevis) dickkopf related protein-1 Dickkopf-1 dickkopf-related protein 1 DKK-1 hDkk-1 SKdickkopf-1 like; dickkopf 1 homolog; dickkopf related protein-1; Dickkopf-1; dickkopf-1 like; dickkopf-like protein 1; Dickkopf-related protein 1; hDkk-1; SK
Gene Aliases: DKK-1; DKK1; SK; UNQ492/PRO1008
UniProt ID: (Human) O94907
Entrez Gene ID: (Human) 22943
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.