Novus Biologicals
Manufacturer Code:NBP132388
Catalog # NBP132388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Dishevelled 2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dishevelled 2 (homologous to Drosophila dsh); dishevelled 2 (homologous to Drosophila dsh) dishevelled dsh homolog 2 (Drosophila) Dishevelled-2 DSH homolog 2 segment polarity protein dishevelled homolog DVL-2; dishevelled, dsh homolog 2; Dishevelled-2; DSH homolog 2; Segment polarity protein dishevelled homolog DVL-2
Gene Aliases: DVL2
UniProt ID: (Human) O14641
Entrez Gene ID: (Human) 1856
Molecular Function: enzyme modulator signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.