Novus Biologicals
Manufacturer Code:NBP270569
Catalog # NBP270569
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6G7 |
Immunogen | Full-length protein expressed in 293T cell transfected with human DHFR expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: DHFRP1 dihydrofolate reductase DYR EC 1.5.1.3; Dihydrofolate reductase
Gene Aliases: DHFR; DHFRP1; DYR
UniProt ID: (Human) P00374
Entrez Gene ID: (Human) 1719
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.