Invitrogen
Manufacturer Code:MA527595
Catalog # PIMA527595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S167-7 |
Immunogen | Fusion protein amino acids 1638-1899 of mouse Endoribonuclease Dicer |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dicer 1 ribonuclease III; dicer 1, double-stranded RNA-specific endoribonuclease; dicer 1, ribonuclease type III; Dicer1 DCR DCR1 DCR-1 Double-strand-specific ribonuclease Helicase with RNase motif HERNA Helicase MOI KIAA0928 Endoribonuclease Dicer; Dicer1, Dcr-1 homolog; double-strand-specific ribonuclease mDCR-1; Endoribonuclease Dicer; Helicase MOI; Helicase with RNase motif
Gene Aliases: 1110006F08Rik; D12Ertd7e; DCR1; DICER; DICER1; Dicer1e; HERNA; K12H4.8-LIKE; KIAA0928; mKIAA0928; MNG1; RMSE2
UniProt ID: (Human) Q9UPY3
Entrez Gene ID: (Human) 23405, (Mouse) 192119, (Rat) 299284
Molecular Function:
DNA binding protein
endodeoxyribonuclease
nuclease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.