Abnova
Manufacturer Code:PAB8674
Catalog # 89-133-844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 308 of mouse Dgat1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Dgat1.
For Research Use Only
Protein Aliases: Acyl-CoA retinol O-fatty-acyltransferase; ARAT; C75990; D15Ertd23e; Dgat; diacylglycerol acyltransferase 1; Diacylglycerol O-acyltransferase 1; Diglyceride acyltransferase; retinol O-fatty-acyltransferase
Gene Aliases: ARAT; C75990; D15Ertd23e; Dgat; Dgat1
UniProt ID: (Mouse) Q9Z2A7
Entrez Gene ID: (Mouse) 13350
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.