Abnova
Manufacturer Code:PAB13074
Catalog # 89-116-327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 314-329 of mouse Dffb. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Dffb.
For Research Use Only
Protein Aliases: 40kDa; 5730477D02Rik; CAD; Caspase-activated deoxyribonuclease; caspase-activated DNase; CPAN; DFF-40; DFF40; Didff; DNA fragmentation factor 40 kDa subunit; DNA fragmentation factor subunit beta; DNA fragmentation factor, 40 kD, beta subunit; DNase inhibited by DNA fragmentation factor
Gene Aliases: 40kDa; 5730477D02Rik; CAD; CPAN; DFF40; Dffb; Didff
UniProt ID: (Mouse) O54788
Entrez Gene ID: (Mouse) 13368
Molecular Function:
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.