Abnova
Manufacturer Code:PAB13071
Catalog # 89-116-324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of mouse Dffa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Dffa.
For Research Use Only
Protein Aliases: A330085O09Rik; DFF-45; DFF35; Dff45; DNA fragmentation factor 45 kDa subunit; DNA fragmentation factor subunit alpha; ICAD; ICAD-L; ICAD-S; Inhibitor of CAD
Gene Aliases: A330085O09Rik; DFF35; Dff45; Dffa; ICAD; ICAD-L; ICAD-S
UniProt ID: (Mouse) O54786
Entrez Gene ID: (Mouse) 13347
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.