R&D Systems
Manufacturer Code:MAB9080
Catalog # MAB9080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 824038 |
Immunogen | <em>E. coli</em>-derived recombinant human Desmoplakin Asn1796-Val2046 Accession # P15924 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 250/210 kDa paraneoplastic pemphigus antigen; 250/210 kDa paraneoplastic pemphigus antigen DCWHKTA desmoplakin desmoplakin (DPI DPII) desmoplakin I desmoplakin II DP DPI DPII DSP KPPS2 PPKS2; Desmoplakin; DP
Gene Aliases: DCWHKTA; DP; DSP
UniProt ID: (Human) P15924
Entrez Gene ID: (Human) 1832
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.